Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001267868.1 | NP_001254797.1 | 9 | 1660 | |
NM_001267869.1 | NP_001254798.1 | 6 | 1347 | ||
NM_001267870.1 | NP_001254799.1 | 5 | 1187 | ||
NM_001267871.1 | NP_001254800.1 | 6 | 1116 | ||
NM_198533.2 | NP_940935.1 | 8 | 1359 | ||
NM_198704.2 | NP_941993.1 | 6 | 1184 | ||
NM_198705.2 | NP_941994.1 | 6 | 1279 | ||
NM_198706.2 | NP_941995.1 | 8 | 1522 | ||
NM_198707.2 | NP_941996.1 | 7 | 1459 | ||
NM_198708.2 | NP_941997.1 | 7 | 1410 | ||
GenBank mRNA | AK308206.1 | - | 9 | 1401 | |
AY268351.1 | - | 6 | 1020 | ||
AY268352.1 | - | 8 | 1223 | ||
AY268353.1 | - | 7 | 1183 | ||
AY268354.1 | - | 7 | 1111 | ||
AY313894.1 | - | 8 | 1088 | ||
AY313895.1 | - | 6 | 925 | ||
AY313896.1 | - | 6 | 857 | ||
BC053546.1 | - | 7 | 1040 | ||
BC072434.1 | - | 6 | 1096 | ||
BC094798.1 | - | 6 | 1103 |