Gene Transcripts
- Gene Symbol
- KCP
- Entrez Gene ID
- 375616
- Gene Name
- kielin/chordin-like protein
- Gene Aliases
- CRIM2, KCP1, NET67
- Chromosome Location
- Chr. 7: 128870645 - 128910719 on Build GRCh38
- UniGene ID
- Hs.371746
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135914.1 |
NP_001129386.1
|
2, 3 |
255 |
|
NM_199349.2 |
NP_955381.2
|
2, 3 |
255 |
|
XM_017012184.1 |
XP_016867673.1
|
2, 3 |
255 |
|
XM_017012185.1 |
XP_016867674.1
|
2, 3 |
255 |
|
XM_017012186.1 |
XP_016867675.1
|
2, 3 |
255 |
|
XM_017012187.1 |
XP_016867676.1
|
2, 3 |
255 |
|
XM_017012188.1 |
XP_016867677.1
|
2, 3 |
255 |
|
XM_017012189.1 |
XP_016867678.1
|
2, 3 |
255 |
|
XM_017012190.1 |
XP_016867679.1
|
2, 3 |
255 |
|
XM_017012193.1 |
XP_016867682.1
|
2, 3 |
255 |
|
XM_017012194.1 |
XP_016867683.1
|
2, 3 |
255 |
|
GenBank mRNA |
AK122706.1 |
- |
2, 3 |
255 |
|
View Full Product Details