Gene Transcripts

Gene Symbol
C2orf78
Entrez Gene ID
388960
Gene Name
chromosome 2 open reading frame 78
Gene Aliases
COG5373, hCG1989538
Chromosome Location
Chr. 2: 73784189 - 73817148 on Build GRCh38
UniGene ID
Hs.456833
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080474.1 NP_001073943.1 3 1255
XM_005264302.3 XP_005264359.1 3 1200
GenBank mRNA AK125975.1 - 3 1256

View Full Product Details