Gene Transcripts

Gene Symbol
PLEKHM3
Entrez Gene ID
389072
Gene Name
pleckstrin homology domain containing M3
Gene Aliases
DAPR, PLEKHM1L
Chromosome Location
Chr. 2: 207821288 - 208025675 on Build GRCh38
UniGene ID
Hs.159188
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080475.2 NP_001073944.1 3 1207
XM_005246545.3 XP_005246602.1 3 1207
XM_006712521.3 XP_006712584.1 3 1207
XM_011511160.2 XP_011509462.1 4 1542
XM_011511163.2 XP_011509465.1 3 1207
XM_017004071.1 XP_016859560.1 3 1207
XM_017004072.1 XP_016859561.1 2 1392
GenBank mRNA BC151137.1 - 2 846
BX648983.1 - 3 1146

View Full Product Details