Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159547.1 | NP_001153019.1 | 5 | 1755 | |
NM_207406.3 | NP_997289.2 | 5 | 1755 | ||
XM_017008185.1 | XP_016863674.1 | 4 | 1412 | ||
GenBank mRNA | AB126828.1 | - | 5 | 1755 | |
AB126829.1 | - | 5 | 1755 | ||
AK125953.1 | - | 5 | 941 | ||
BC128592.1 | - | 5 | 840 | 40127911 | |
BC128593.1 | - | 5 | 840 | 40127915 |