Gene Transcripts

Gene Symbol
OR6Y1
Entrez Gene ID
391112
Gene Name
olfactory receptor family 6 subfamily Y member 1
Gene Aliases
OR1-11, OR6Y2
Chromosome Location
Chr. 1: 158547128 - 158548105 on Build GRCh38
UniGene ID
Hs.553780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005189.1 NP_001005189.1 1 638

View Full Product Details