Gene Transcripts
- Gene Symbol
- GALC
- Entrez Gene ID
- 2581
- Gene Name
- galactosylceramidase
- Gene Aliases
- Chromosome Location
- Chr. 14: 87933014 - 87993665 on Build GRCh38
- UniGene ID
- Hs.513439
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000153.3 |
NP_000144.2
|
16, 17 |
2011 |
|
NM_001201401.1 |
NP_001188330.1
|
15, 16 |
1942 |
|
NM_001201402.1 |
NP_001188331.1
|
16, 17 |
1992 |
|
XM_011536618.2 |
XP_011534920.1
|
16, 17 |
2293 |
|
GenBank mRNA |
AK294040.1 |
- |
17, 18 |
1856 |
|
AK296530.1 |
- |
15, 16 |
1833 |
|
AK302956.1 |
- |
16, 17 |
1831 |
|
BC036518.2 |
- |
16, 17 |
1863 |
5267705 |
D25283.1 |
- |
16, 17 |
1894 |
|
H80485.1 |
- |
3, 4 |
267 |
|
L23116.1 |
- |
16, 17 |
1903 |
|
View Full Product Details