Gene Transcripts

Gene Symbol
GDF3
Entrez Gene ID
9573
Gene Name
growth differentiation factor 3
Gene Aliases
KFS3, MCOP7, MCOPCB6
Chromosome Location
Chr. 12: 7689785 - 7695764 on Build GRCh38
UniGene ID
Hs.86232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020634.1 NP_065685.1 1 186
GenBank mRNA AF110641.1 - 1 164
AF263538.1 - 1 186
AY358875.1 - 1 215
BC030959.1 - 1 198 4610141

View Full Product Details