Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004629.1 | NP_004620.1 | 8 | 1457 | |
GenBank mRNA | AJ007669.1 | - | 8 | 1457 | |
AK293427.1 | - | 9 | 1147 | ||
AK311348.1 | - | 7 | 931 | ||
AK312987.1 | - | 8 | 1382 | ||
BC000032.2 | - | 8 | 1103 | 3507258 | |
BC011623.2 | - | 8 | 1230 | 4053144 | |
BT009813.1 | - | 8 | 965 | ||
KU159422.1 | - | 8 | 1488 | ||
KU178053.1 | - | 8 | 965 | ||
KU178054.1 | - | 8 | 965 | ||
KU178055.1 | - | 8 | 965 | ||
KU178056.1 | - | 5 | 539 | ||
U70310.1 | - | 8 | 1099 |