Gene Transcripts

Gene Symbol
MOCS1
Entrez Gene ID
4337
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
MIG11, MOCOD, MOCODA
Chromosome Location
Chr. 6: 39904258 - 39934551 on Build GRCh38
UniGene ID
Hs.357128
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001075098.3 NP_001068566.1 11 2652
NM_005943.5 NP_005934.2 10 2656
NR_033233.1 9 2462
XM_011514632.2 XP_011512934.1 11 2548
XM_011514633.2 XP_011512935.1 10 2500
XM_011514634.2 XP_011512936.1 10 2445
XM_017010888.1 XP_016866377.1 10 2470
GenBank mRNA AF034374.1 - 11 2535
AJ224328.1 - 10 2653
AK056740.1 - 9 2408
AK091306.1 - 10 2384

View Full Product Details