Gene Transcripts
- Gene Symbol
- AGK
- Entrez Gene ID
- 55750
- Gene Name
- acylglycerol kinase
- Gene Aliases
- CATC5, CTRCT38, MTDPS10, MULK
- Chromosome Location
- Chr. 7: 141551278 - 141655244 on Build GRCh38
- UniGene ID
- Hs.743318
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018238.3 |
NP_060708.1
|
5, 6 |
463 |
|
XM_005250023.3 |
XP_005250080.1
|
5, 6 |
359 |
|
XM_011516397.2 |
XP_011514699.1
|
5, 6 |
929 |
|
GenBank mRNA |
AJ278150.1 |
- |
4, 5 |
319 |
|
AK001704.1 |
- |
5, 6 |
331 |
|
AK222769.1 |
- |
5, 6 |
325 |
|
AK299131.1 |
- |
5, 6 |
322 |
|
AK304502.1 |
- |
6, 7 |
350 |
|
BC022777.1 |
- |
4, 5 |
317 |
3913448 |
DB032353.1 |
- |
5, 6 |
463 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM547306.1 |
- |
|
743 |
|
Chr. - - [-] |
View Full Product Details