Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000939.3 | NP_000930.1 | 3 | 1352 | |
NM_001035256.2 | NP_001030333.1 | 4 | 1402 | ||
NM_001319204.1 | NP_001306133.1 | 4 | 1432 | ||
NM_001319205.1 | NP_001306134.1 | 3 | 1382 | ||
GenBank mRNA | AW951218.1 | - | 135 | ||
T29537.1 | - | 1 | 135 |