Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000133.3 | NP_000124.1 | 2 | 172 | |
NM_001313913.1 | NP_001300842.1 | 2 | 172 | ||
XM_005262397.4 | XP_005262454.1 | 2 | 188 | ||
GenBank mRNA | A13997.1 | - | 2 | 144 | |
AB186358.1 | - | 2 | 143 | ||
AK292749.1 | - | 2 | 143 | ||
BC109214.1 | - | 2 | 172 | 40007636 | |
BC109215.1 | - | 2 | 172 | 40007637 | |
FR846238.1 | - | 2 | 143 | ||
FR846239.1 | - | 2 | 143 | ||
FR846240.1 | - | 2 | 143 | ||
J00136.1 | - | 2 | 143 | ||
J00137.1 | - | 2 | 144 | ||
M11309.1 | - | 2 | 143 | ||
M35672.1 | - | 1 | 38 |