Gene Transcripts

Gene Symbol
COL11A2
Entrez Gene ID
1302
Gene Name
collagen type XI alpha 2
Gene Aliases
DFNA13, DFNB53, FBCG2, HKE5, PARP, STL3
Chromosome Location
Chr. 6: 33162692 - 33193009 on Build GRCh38
UniGene ID
Hs.390171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080680.2 NP_542411.2 8 1261
XM_011514299.2 XP_011512601.1 3 552
XM_017010250.1 XP_016865739.1 9 1526

View Full Product Details