Gene Transcripts

Gene Symbol
RBM10
Entrez Gene ID
8241
Gene Name
RNA binding motif protein 10
Gene Aliases
DXS8237E, GPATC9, GPATCH9, S1-1, TARPS, ZRANB5
Chromosome Location
Chr. X: 47145196 - 47186815 on Build GRCh38
UniGene ID
Hs.401509
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204466.1 NP_001191395.1 10 1291
NM_001204467.1 NP_001191396.1 11 1519
NM_001204468.1 NP_001191397.1 11 1553
NM_005676.4 NP_005667.2 11 1522
NM_152856.2 NP_690595.1 10 1288
XM_005272677.4 XP_005272734.1 11 1572
XM_005272678.4 XP_005272735.1 10 1106
XM_005272679.4 XP_005272736.1 10 1103
XM_006724563.1 XP_006724626.1 7 739
XM_011543989.2 XP_011542291.1 3 463
XM_017029884.1 XP_016885373.1 10 1442
XM_017029885.1 XP_016885374.1 10 1214
GenBank mRNA AK292758.1 - 10 1242
AL137421.1 - 12 1411
BC000681.2 - 8 744
BC003089.1 - 10 1227 3536058
BC004181.2 - 11 1475 2961094
BC008733.2 - 11 1507 3163064
BC024153.2 - 10 1137 3845258
BX537969.1 - 11 1504
D50912.1 - 11 1370
KU178411.1 - 9 895
KU178412.1 - 10 1129

View Full Product Details