Gene Transcripts

Gene Symbol
C6
Entrez Gene ID
729
Gene Name
complement component 6
Gene Aliases
Chromosome Location
Chr. 5: 41142116 - 41261486 on Build GRCh38
UniGene ID
Hs.481992
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000065.3 NP_000056.2 9 1457
NM_001115131.2 NP_001108603.2 9 1484
XM_005248357.2 XP_005248414.1 9 1532
XM_006714496.3 XP_006714559.1 9 1532
XM_011514114.2 XP_011512416.1 9 1532
XM_011514115.2 XP_011512417.1 9 1453
XM_011514116.2 XP_011512418.1 10 1569
XM_011514117.2 XP_011512419.1 10 1611
XM_011514118.2 XP_011512420.1 9 1349
XM_011514119.2 XP_011512421.1 9 1532
XM_011514121.2 XP_011512423.1 4 618
XM_017009818.1 XP_016865307.1 10 1592
XM_017009819.1 XP_016865308.1 5 795
GenBank mRNA AB126592.1 - 8 1219
AK292509.1 - 9 1355
AK313673.1 - 9 1376
BC035723.1 - 9 1484 5591446
J04506.1 - 9 1285
J05024.1 - 9 1276
J05064.1 - 9 1374

View Full Product Details