Gene Transcripts

Gene Symbol
RPS24
Entrez Gene ID
6229
Gene Name
ribosomal protein S24
Gene Aliases
DBA3, S24
Chromosome Location
Chr. 10: 78033760 - 78056813 on Build GRCh38
UniGene ID
Hs.280130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001026.4 NP_001017.1 1, 2 152
NM_001142282.1 NP_001135754.1 1, 2 152
NM_001142283.1 NP_001135755.1 1, 2 152
NM_001142284.1 NP_001135756.1 1, 2 152
NM_001142285.1 NP_001135757.1 1, 2 152
NM_033022.3 NP_148982.1 1, 2 152
GenBank mRNA AF147330.1 - 1, 2 33
AK311776.1 - 1, 2 49
AK311910.1 - 1, 2 49
BC000523.2 - 1, 2 29 2961125
BC003149.1 - 1, 2 25
BC071926.1 - 1, 2 34 5556377
BG610671.1 - 96
BG944046.1 - 1, 2 153
BM745909.1 - 1, 2 49
BX537975.1 - 1, 2 48
BX647260.1 - 1, 2 38
CD176901.1 - 1, 2 75
CD677556.1 - 1, 2 28
DB116225.1 - 1, 2 49
M31520.1 - 1, 2 152

View Full Product Details