Gene Transcripts

Gene Symbol
RAB19
Entrez Gene ID
401409
Gene Name
RAB19, member RAS oncogene family
Gene Aliases
RAB19B
Chromosome Location
Chr. 7: 140403740 - 140426881 on Build GRCh38
UniGene ID
Hs.583545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008749.2 NP_001008749.2 3 538
XM_005249996.3 XP_005250053.1 3 821
XM_005249997.3 XP_005250054.1 3 804
XM_005249998.3 XP_005250055.1 3 799
XM_017012207.1 XP_016867696.1 3 734
GenBank mRNA BC140796.1 - 2 408
DN996121.1 - 3 512

View Full Product Details