Gene Transcripts

Gene Symbol
OR2T2
Entrez Gene ID
401992
Gene Name
olfactory receptor family 2 subfamily T member 2
Gene Aliases
OR1-43, OR2T2P
Chromosome Location
Chr. 1: 248452798 - 248453772 on Build GRCh38
UniGene ID
Hs.554482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004136.1 NP_001004136.1 1 525
GenBank mRNA BC136961.1 - 1 547
BC136962.1 - 1 547 9020954

Gene Symbol
OR2T35
Entrez Gene ID
403244
Gene Name
olfactory receptor family 2 subfamily T member 35
Gene Aliases
Chromosome Location
Chr. 1: 248638287 - 248639258 on Build GRCh38
UniGene ID
Hs.534846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001827.1 NP_001001827.1 1 522

View Full Product Details