Gene Transcripts

Gene Symbol
AARD
Entrez Gene ID
441376
Gene Name
alanine and arginine rich domain containing protein
Gene Aliases
C8orf85
Chromosome Location
Chr. 8: 116937960 - 116944000 on Build GRCh38
UniGene ID
Hs.437551
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025357.2 NP_001020528.1 2 437
XM_011517043.2 XP_011515345.1 2 815
GenBank mRNA AB181519.1 - 2 418
BC140924.1 - 2 463 9056736
BC142621.1 - 2 437 40147448

View Full Product Details