Gene Transcripts
- Gene Symbol
- ANKRD20A2
- Entrez Gene ID
- 441430
- Gene Name
- ankyrin repeat domain 20 family member A2
- Gene Aliases
- Chromosome Location
- Chr. 9: 40223285 - 40282360 on Build GRCh38
- UniGene ID
- Hs.632663
- Species
- Homo sapiens
- Gene Symbol
- ANKRD20A1
- Entrez Gene ID
- 84210
- Gene Name
- ankyrin repeat domain 20 family member A1
- Gene Aliases
- ANKRD20A
- Chromosome Location
- Chr. 9: 67859315 - 67902394 on Build GRCh38
- UniGene ID
- Hs.632671
- Species
- Homo sapiens
- Gene Symbol
- ANKRD20A3
- Entrez Gene ID
- 441425
- Gene Name
- ankyrin repeat domain 20 family member A3
- Gene Aliases
- Chromosome Location
- Chr. 9: 66093946 - 66153193 on Build GRCh38
- UniGene ID
- Hs.632663
- Species
- Homo sapiens
- Gene Symbol
- ANKRD20A4
- Entrez Gene ID
- 728747
- Gene Name
- ankyrin repeat domain 20 family member A4
- Gene Aliases
- Chromosome Location
- Chr. 9: 64369563 - 64412691 on Build GRCh38
- UniGene ID
- Hs.645502
- Species
- Homo sapiens
- Gene Symbol
- ANKRD20A8P
- Entrez Gene ID
- 729171
- Gene Name
- ankyrin repeat domain 20 family member A8, pseudogene
- Gene Aliases
- ANKRD20B
- Chromosome Location
- Chr. 2: 94760928 - 94857075 on Build GRCh38
- UniGene ID
- Hs.456852
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003366.2 |
|
5 |
523 |
|
View Full Product Details