Gene Transcripts

Gene Symbol
GAB1
Entrez Gene ID
2549
Gene Name
GRB2 associated binding protein 1
Gene Aliases
Chromosome Location
Chr. 4: 143336830 - 143474565 on Build GRCh38
UniGene ID
Hs.618456
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002039.3 NP_002030.2 4 1269
NM_207123.2 NP_997006.1 4 1269
XM_006714167.3 XP_006714230.1 5 2167
XM_006714168.3 XP_006714231.1 5 1831
XM_017007966.1 XP_016863455.1 6 7994
XM_017007967.1 XP_016863456.1 5 1079
XM_017007968.1 XP_016863457.1 5 7911
XM_017007969.1 XP_016863458.1 5 3243
XM_017007970.1 XP_016863459.1 5 2998
XM_017007971.1 XP_016863460.1 4 2917
GenBank mRNA AK289767.1 - 4 1212
AK295684.1 - 4 913
AK314635.1 - 4 949
BC064848.1 - 4 1133 30386955
U43885.1 - 4 1031

View Full Product Details