Gene Transcripts
- Gene Symbol
- TMEM14EP
- Entrez Gene ID
- 645843
- Gene Name
- transmembrane protein 14E, pseudogene
- Gene Aliases
- TMEM14E
- Chromosome Location
- Chr. 3: 152339698 - 152340990 on Build GRCh38
- UniGene ID
- Hs.632562
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_132656.1 |
|
1 |
307 |
|
GenBank mRNA |
AW866456.1 |
- |
1 |
34 |
|
AY358260.1 |
- |
1 |
307 |
|
View Full Product Details