Gene Transcripts
- Gene Symbol
- RPS16P5
- Entrez Gene ID
- 647190
- Gene Name
- ribosomal protein S16 pseudogene 5
- Gene Aliases
- RPS16_3_702
- Chromosome Location
- Chr. 6: 53334460 - 53337617 on Build GRCh38
- UniGene ID
- Hs.520189
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046241.1 |
|
1 |
537 |
|
GenBank mRNA |
AK097937.2 |
- |
1 |
537 |
|
View Full Product Details