Gene Transcripts

Gene Symbol
OR4N2
Entrez Gene ID
390429
Gene Name
olfactory receptor family 4 subfamily N member 2
Gene Aliases
OR14-13, OR14-8
Chromosome Location
Chr. 14: 19803770 - 19828372 on Build GRCh38
UniGene ID
Hs.512490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004723.2 NP_001004723.1 2 749

Gene Symbol
OR4N3P
Entrez Gene ID
390539
Gene Name
olfactory receptor family 4 subfamily N member 3 pseudogene
Gene Aliases
OR15-2
Chromosome Location
Chr. 15: 22125511 - 22126447 on Build GRCh38
UniGene ID
Hs.666555
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028067.1 1 665
GenBank mRNA BX114189.1 - 1 211

View Full Product Details