Gene Transcripts

Gene Symbol
MAP2K3
Entrez Gene ID
5606
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
Chromosome Location
Chr. 17: 21284656 - 21315240 on Build GRCh38
UniGene ID
Hs.514012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316332.1 NP_001303261.1 13 1847
NM_002756.4 NP_002747.2 12 1464
NM_145109.2 NP_659731.1 12 1722
XM_005256723.2 XP_005256780.1 13 1762
XM_011523958.2 XP_011522260.1 11 1381
XM_011523959.1 XP_011522261.1 14 1729
XM_017024857.1 XP_016880346.1 12 1591
XM_017024858.1 XP_016880347.1 13 1579
XM_017024859.1 XP_016880348.1 14 1919
GenBank mRNA AK093838.1 - 13 1766
BC032478.1 - 12 1657 5215093
D87116.1 - 12 1514
L36719.1 - 14 1707
U66839.1 - 12 1583

Gene Symbol
LOC100996792
Entrez Gene ID
100996792
Gene Name
dual specificity mitogen-activated protein kinase kinase 3
Gene Aliases
Chromosome Location
Chr. 17: 21772912 - 21783444 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011524134.2 XP_011522436.2 5 828

View Full Product Details