Gene Transcripts

Gene Symbol
GGT1
Entrez Gene ID
2678
Gene Name
gamma-glutamyltransferase 1
Gene Aliases
CD224, D22S672, D22S732, GGT, GGT 1, GTG
Chromosome Location
Chr. 22: 24583750 - 24629005 on Build GRCh38
UniGene ID
Hs.595809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288833.1 NP_001275762.1 5 896
NM_013421.2 NP_038265.2 6 824
NM_013430.2 NP_038347.2 5 584
GenBank mRNA AL832738.1 - 800
BC025927.1 - 3 464 4548861
BU157907.1 - 5 567
BU164348.1 - 5 591
CR456494.1 - 2 174
J04131.1 - 6 766
J05235.1 - 4 841
M24087.1 - 1 99
M24903.1 - 5 584
X60069.1 - 3 455

Gene Symbol
GGT2
Entrez Gene ID
728441
Gene Name
gamma-glutamyltransferase 2
Gene Aliases
GGT, GGT 2
Chromosome Location
Chr. 22: 21207972 - 21282982 on Build GRCh38
UniGene ID
Hs.568255
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006724392.3 XP_006724455.1 4 647
XM_017029179.1 XP_016884668.1 5 1328
XM_017029180.1 XP_016884669.1 4 786
XM_017029181.1 XP_016884670.1 4 1224
XM_017029182.1 XP_016884671.1 5 1328
XM_017029185.1 XP_016884674.1 5 808
XM_017029186.1 XP_016884675.1 7 1487

Gene Symbol
GGT3P
Entrez Gene ID
2679
Gene Name
gamma-glutamyltransferase 3 pseudogene
Gene Aliases
GGT3
Chromosome Location
Chr. 22: 18773689 - 18791961 on Build GRCh38
UniGene ID
Hs.595809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003267.1 2 293
GenBank mRNA BC143607.1 - 2 159

Gene Symbol
GGT8P
Entrez Gene ID
645367
Gene Name
gamma-glutamyltransferase 8 pseudogene
Gene Aliases
GGT1P1
Chromosome Location
Chr. 2: 91775342 - 91782127 on Build GRCh38
UniGene ID
Hs.650223
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003503.1 1 699
GenBank mRNA AK055838.1 - 1 699

Gene Symbol
LOC102724197
Entrez Gene ID
102724197
Gene Name
inactive gamma-glutamyltranspeptidase 2-like
Gene Aliases
GGT2
Chromosome Location
Chr. 22|NT_187386: 65299 - 86956 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006724921.3 XP_006724984.1 3 932
XM_006724924.3 XP_006724987.1 3 932
XM_011546256.2 XP_011544558.1 3 935
XM_017030133.1 XP_016885622.1 4 497
XM_017030134.1 XP_016885623.1 4 500
XM_017030135.1 XP_016885624.1 4 925
XM_017030136.1 XP_016885625.1 5 967
XM_017030137.1 XP_016885626.1 5 964
XM_017030138.1 XP_016885627.1 5 487
XM_017030139.1 XP_016885628.1 5 490
XM_017030140.1 XP_016885629.1 6 957
XM_017030141.1 XP_016885630.1 6 954
XM_017030142.1 XP_016885631.1 4 798
XM_017030143.1 XP_016885632.1 3 933
XM_017030145.1 XP_016885634.1 3 932
XM_017030146.1 XP_016885635.1 3 949

Gene Symbol
LOC102724823
Entrez Gene ID
102724823
Gene Name
inactive gamma-glutamyltranspeptidase 2
Gene Aliases
Chromosome Location
Chr. 22|NT_187389: 6 - 18670 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017030162.1 XP_016885651.1 4 966

View Full Product Details