Gene Transcripts

Gene Symbol
RPL7
Entrez Gene ID
6129
Gene Name
ribosomal protein L7
Gene Aliases
L7, humL7-1
Chromosome Location
Chr. 8: 73290639 - 73294494 on Build GRCh38
UniGene ID
Hs.421257
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000971.3 NP_000962.2 1 208
XM_006716463.2 XP_006716526.1 3 801
GenBank mRNA AK225246.1 - 1 207
AK225254.1 - 1 206
AK291119.1 - 3 206
AK313365.1 - 3 206
BC006095.1 - 1 196 2821364
BC008850.2 - 1 233 4138113
BC009599.1 - 1 208 4134557
BC071671.1 - 1 189 4392472
BC071894.1 - 1 193 2906310
BC071895.1 - 1 191 6568654
BC087837.1 - 1 192 6170371
CR456773.1 - 1 186
L16558.1 - 1 186
X52967.1 - 3 207
X57958.1 - 1 223
X57959.1 - 1 196

View Full Product Details