Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001097622.1 | NP_001091091.1 | 3 | 333 | |
XM_011515485.2 | XP_011513787.1 | 4 | 280 | ||
GenBank mRNA | BC069468.1 | - | 3 | 241 | |
BC069512.1 | - | 3 | 241 | 7262741 | |
BC069550.1 | - | 3 | 241 | 7262753 | |
BC113704.1 | - | 3 | 333 | 8322756 | |
BC113706.1 | - | 3 | 333 | 8322758 | |
BC147028.1 | - | 3 | 335 | 9056956 | |
BC147029.1 | - | 3 | 335 | 9056957 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006188.3 | NP_006179.2 | 3 | 241 |