Gene Transcripts

Gene Symbol
GLP1R
Entrez Gene ID
2740
Gene Name
glucagon like peptide 1 receptor
Gene Aliases
GLP-1, GLP-1-R, GLP-1R
Chromosome Location
Chr. 6: 39048781 - 39091303 on Build GRCh38
UniGene ID
Hs.351883
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002062.4 NP_002053.3 11 1210
NR_136562.1 11 1210
NR_136563.1 11 1210
XM_017010750.1 XP_016866239.1 10 1745
GenBank mRNA BC112126.1 - 11 1222
BC113493.1 - 11 1210 8322545
KR138540.1 - 11 1176
L23503.1 - 11 1175
U01104.1 - 11 1192
U01156.1 - 11 1150
U01157.1 - 11 1150
U10037.1 - 11 1276

View Full Product Details