Gene Transcripts

Gene Symbol
NUS1
Entrez Gene ID
116150
Gene Name
NUS1 dehydrodolichyl diphosphate synthase subunit
Gene Aliases
C6orf68, MGC:7199, NgBR, TANGO14
Chromosome Location
Chr. 6: 117675454 - 117710727 on Build GRCh38
UniGene ID
Hs.289008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138459.3 NP_612468.1 5 1698
GenBank mRNA AK125119.1 - 5 1660
AK126335.1 - 6 1412
AK130093.1 - 4 1015
BC013026.2 - 5 1645 3531630
BC063794.1 - 5 1652 6500832
BC066910.1 - 5 1692 4830860
BC110325.1 - 5 1654 5553827

View Full Product Details