Gene Transcripts

Gene Symbol
PRR20A
Entrez Gene ID
122183
Gene Name
proline rich 20A
Gene Aliases
PRR20
Chromosome Location
Chr. 13: 57140918 - 57143939 on Build GRCh38
UniGene ID
Hs.525056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198441.2 NP_940843.1 3 1087
GenBank mRNA AK097615.1 - 3 1087
BC120931.1 - 3 956 40114895
BC120932.1 - 3 956 40114901

Gene Symbol
PRR20B
Entrez Gene ID
729233
Gene Name
proline rich 20B
Gene Aliases
Chromosome Location
Chr. 13: 57147488 - 57150509 on Build GRCh38
UniGene ID
Hs.723090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130404.1 NP_001123876.1 3 1087

Gene Symbol
PRR20C
Entrez Gene ID
729240
Gene Name
proline rich 20C
Gene Aliases
Chromosome Location
Chr. 13: 57154061 - 57157082 on Build GRCh38
UniGene ID
Hs.525056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130405.1 NP_001123877.1 3 1087

Gene Symbol
PRR20D
Entrez Gene ID
729246
Gene Name
proline rich 20D
Gene Aliases
Chromosome Location
Chr. 13: 57160632 - 57163653 on Build GRCh38
UniGene ID
Hs.525056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130406.1 NP_001123878.1 3 1087

Gene Symbol
PRR20E
Entrez Gene ID
729250
Gene Name
proline rich 20E
Gene Aliases
Chromosome Location
Chr. 13: 57167197 - 57170218 on Build GRCh38
UniGene ID
Hs.525056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130407.1 NP_001123879.1 3 1087

View Full Product Details