Gene Transcripts
- Gene Symbol
- Adam5
- Entrez Gene ID
- 11499
- Gene Name
- a disintegrin and metallopeptidase domain 5
- Gene Aliases
- tMDC II, tMDCII
- Chromosome Location
- Chr. 8: 24727093 - 24824369 on Build GRCm38
- UniGene ID
- Mm.143889
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001272057.1 |
NP_001258986.1
|
10, 11 |
1276 |
|
NM_001272058.1 |
NP_001258987.1
|
10, 11 |
1276 |
|
NM_001272059.1 |
NP_001258988.1
|
10, 11 |
1276 |
|
NM_007401.3 |
NP_031427.2
|
10, 11 |
1276 |
|
GenBank mRNA |
AK016574.1 |
- |
10, 11 |
932 |
|
AK029519.1 |
- |
10, 11 |
966 |
|
AK161262.1 |
- |
10, 11 |
1276 |
|
AK161415.1 |
- |
7, 8 |
654 |
|
AK161417.1 |
- |
10, 11 |
946 |
|
BC094237.1 |
- |
10, 11 |
973 |
30604517 |
U22059.1 |
- |
10, 11 |
940 |
|
View Full Product Details