Gene Transcripts

Gene Symbol
Aire
Entrez Gene ID
11634
Gene Name
autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
Gene Aliases
Chromosome Location
Chr. 10: 78030022 - 78043610 on Build GRCm38
UniGene ID
Mm.35300
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271549.1 NP_001258478.1 6, 7 890
NM_001271552.1 NP_001258481.1 6, 7 890
NM_001271553.1 NP_001258482.1 6, 7 890
NM_001271556.1 NP_001258485.1 6, 7 890
NM_001271557.1 NP_001258486.1 6, 7 890
NM_009646.2 NP_033776.1 6, 7 890
GenBank mRNA AF079536.1 - 6, 7 860
AF128115.1 - 6, 7 860
AF128118.1 - 6, 7 860
AF128119.1 - 6, 7 860
AF128122.1 - 6, 7 860
AF128123.1 - 6, 7 860
AF128772.1 - 6, 7 803
AF128773.1 - 6, 7 803
AJ132243.1 - 6, 7 803
AJ243821.1 - 6, 7 803
AK153690.1 - 7, 8 944
BC103510.1 - 6, 7 852 40039055
BC103511.1 - 6, 7 852 40039054
BC103518.1 - 6, 7 852 40039058
EU625343.3 - 6, 7 820
JF267399.1 - 6, 7 820

View Full Product Details