Gene Transcripts
- Gene Symbol
- Ctsd
- Entrez Gene ID
- 13033
- Gene Name
- cathepsin D
- Gene Aliases
- CD, CatD
- Chromosome Location
- Chr. 7: 142375916 - 142387870 on Build GRCm38
- UniGene ID
- Mm.231395
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009983.2 |
NP_034113.1
|
4, 5 |
538 |
|
GenBank mRNA |
AK089294.1 |
- |
4, 5 |
496 |
|
AK150363.1 |
- |
4, 5 |
499 |
|
AK150364.1 |
- |
4, 5 |
496 |
|
AK150579.1 |
- |
4, 5 |
496 |
|
AK150622.1 |
- |
4, 5 |
496 |
|
AK150636.1 |
- |
4, 5 |
496 |
|
AK150694.1 |
- |
4, 5 |
496 |
|
AK150768.1 |
- |
4, 5 |
496 |
|
AK150946.1 |
- |
4, 5 |
548 |
|
AK151256.1 |
- |
4, 5 |
496 |
|
AK151275.1 |
- |
4, 5 |
496 |
|
AK151276.1 |
- |
4, 5 |
496 |
|
AK151374.1 |
- |
4, 5 |
496 |
|
AK151829.1 |
- |
4, 5 |
496 |
|
AK151962.1 |
- |
4, 5 |
499 |
|
AK152036.1 |
- |
4, 5 |
509 |
|
AK152045.1 |
- |
4, 5 |
549 |
|
AK152168.1 |
- |
4, 5 |
496 |
|
AK152187.1 |
- |
4, 5 |
496 |
|
AK152188.1 |
- |
4, 5 |
496 |
|
AK152406.1 |
- |
4, 5 |
499 |
|
AK152458.1 |
- |
4, 5 |
496 |
|
AK152567.1 |
- |
4, 5 |
496 |
|
AK152578.1 |
- |
4, 5 |
496 |
|
AK152593.1 |
- |
4, 5 |
526 |
|
AK152638.1 |
- |
4, 5 |
526 |
|
AK152680.1 |
- |
4, 5 |
496 |
|
AK152828.1 |
- |
4, 5 |
496 |
|
AK152881.1 |
- |
4, 5 |
496 |
|
AK152892.1 |
- |
4, 5 |
496 |
|
AK153191.1 |
- |
4, 5 |
496 |
|
AK153291.1 |
- |
4, 5 |
496 |
|
AK159097.1 |
- |
4, 5 |
496 |
|
AK159209.1 |
- |
4, 5 |
496 |
|
AK159440.1 |
- |
4, 5 |
549 |
|
AK159490.1 |
- |
4, 5 |
496 |
|
AK159512.1 |
- |
4, 5 |
538 |
|
AK159580.1 |
- |
4, 5 |
496 |
|
AK159642.1 |
- |
2, 3 |
248 |
|
AK159743.1 |
- |
4, 5 |
500 |
|
AK159896.1 |
- |
4, 5 |
496 |
|
AK159970.1 |
- |
4, 5 |
549 |
|
AK165227.1 |
- |
4, 5 |
566 |
|
AK166864.1 |
- |
4, 5 |
509 |
|
AK166938.1 |
- |
4, 5 |
543 |
|
AK167143.1 |
- |
4, 5 |
534 |
|
AK167366.1 |
- |
4, 5 |
568 |
|
AK167698.1 |
- |
4, 5 |
495 |
|
AK167750.1 |
- |
4, 5 |
509 |
|
AK167867.1 |
- |
4, 5 |
497 |
|
AK169139.1 |
- |
4, 5 |
549 |
|
AK171195.1 |
- |
4, 5 |
497 |
|
AK172583.1 |
- |
4, 5 |
496 |
|
AK194256.1 |
- |
1, 2 |
66 |
|
AK194597.1 |
- |
1, 2 |
67 |
|
AK213259.1 |
- |
1, 2 |
66 |
|
AK213518.1 |
- |
1, 2 |
66 |
|
BC054758.1 |
- |
4, 5 |
568 |
6413315 |
BC057931.1 |
- |
4, 5 |
529 |
5253319 |
X52886.1 |
- |
4, 5 |
538 |
|
X53337.1 |
- |
4, 5 |
478 |
|
View Full Product Details