Gene Transcripts

Gene Symbol
Cyp2d9
Entrez Gene ID
13105
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 9
Gene Aliases
Cyp2d, P450-2D
Chromosome Location
Chr. 15: 82452377 - 82456827 on Build GRCm38
UniGene ID
Mm.226708
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010006.2 NP_034136.2 4 719
GenBank mRNA AK143963.1 - 4 719
BC010593.1 - 4 694 3964178
BC094015.1 - 4 702 5050233
M23997.1 - 4 810
M23998.1 - 4 715
M27168.1 - 4 694

View Full Product Details