Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172129.1 | NP_001165600.1 | 6 | 757 | |
NM_001172130.1 | NP_001165601.1 | 6 | 754 | ||
NM_001172131.1 | NP_001165602.1 | 6 | 754 | ||
NM_001172132.1 | NP_001165603.1 | 7 | 823 | ||
NM_001172133.1 | NP_001165604.1 | 7 | 821 | ||
NM_002110.3 | NP_002101.2 | 6 | 757 | ||
GenBank mRNA | AK026432.1 | - | 6 | 691 | |
AK225819.1 | - | 6 | 703 | ||
AK289896.1 | - | 6 | 745 | ||
AK290928.1 | - | 7 | 769 | ||
AK298726.1 | - | 7 | 767 | ||
AK314335.1 | - | 6 | 670 | ||
BC014435.1 | - | 6 | 643 | 4855747 | |
BC094847.1 | - | 6 | 654 | 5923744 | |
BC108930.1 | - | 6 | 684 | 40025497 | |
BC108931.1 | - | 6 | 684 | 40025500 | |
BC113854.1 | - | 6 | 681 | 40025503 | |
BC114463.1 | - | 6 | 684 | 40025502 | |
M16592.1 | - | 5 | 532 |