Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162425.1 | NP_001155897.1 | 5 | 603 | |
NM_010107.4 | NP_034237.3 | 5 | 639 | ||
GenBank mRNA | AK009144.1 | - | 5 | 636 | |
AK079266.1 | - | 5 | 606 | ||
BC002046.1 | - | 5 | 604 | 3484518 | |
U26188.1 | - | 5 | 598 | ||
U90662.1 | - | 5 | 538 |