Gene Transcripts
- Gene Symbol
- Fcer2a
- Entrez Gene ID
- 14128
- Gene Name
- Fc receptor, IgE, low affinity II, alpha polypeptide
- Gene Aliases
- CD23, Fce2, Fcer2, Ly-42
- Chromosome Location
- Chr. 8: 3680726 - 3694174 on Build GRCm38
- UniGene ID
- Mm.1233
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253737.1 |
NP_001240666.1
|
8, 9 |
577 |
|
NM_001253739.1 |
NP_001240668.1
|
8, 9 |
588 |
|
NM_001253743.1 |
NP_001240672.1
|
9, 10 |
637 |
|
NM_001253745.1 |
NP_001240674.1
|
7, 8 |
514 |
|
NM_001253746.1 |
NP_001240675.1
|
7, 8 |
514 |
|
NM_001253747.1 |
NP_001240676.1
|
8, 9 |
598 |
|
NM_013517.4 |
NP_038545.1
|
9, 10 |
718 |
|
GenBank mRNA |
AK156654.1 |
- |
9, 10 |
721 |
|
AK172591.1 |
- |
9, 10 |
721 |
|
AY069980.1 |
- |
7, 8 |
480 |
|
AY069981.1 |
- |
7, 8 |
480 |
|
BC119164.1 |
- |
8, 9 |
585 |
8733913 |
BC119190.1 |
- |
8, 9 |
585 |
8733939 |
M34163.1 |
- |
9, 10 |
720 |
|
M99371.1 |
- |
9, 10 |
710 |
|
S67072.1 |
- |
8, 9 |
569 |
|
S67073.1 |
- |
7, 8 |
449 |
|
S67074.1 |
- |
8, 9 |
488 |
|
X64223.1 |
- |
8, 9 |
577 |
|
X64224.1 |
- |
8, 9 |
588 |
|
View Full Product Details