Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159544.1 | NP_001153016.1 | 1 | 927 | |
NM_010237.3 | NP_034367.2 | 2 | 771 | ||
GenBank mRNA | AK028855.1 | - | 2 | 711 | |
AK052614.1 | - | 2 | 643 | ||
AK077264.1 | - | 1 | 796 | ||
AK085191.1 | - | 1 | 772 | ||
AK166379.1 | - | 2 | 771 | ||
BC007137.1 | - | 2 | 784 | 3601246 | |
L36132.1 | - | 1 | 806 | ||
Z48757.1 | - | 2 | 636 |