Gene Transcripts

Gene Symbol
Gabrb3
Entrez Gene ID
14402
Gene Name
gamma-aminobutyric acid (GABA) A receptor, subunit beta 3
Gene Aliases
A230092K12Rik, AW049585, Cp1, Gabrb-3, beta3
Chromosome Location
Chr. 7: 57590518 - 57828802 on Build GRCm38
UniGene ID
Mm.8004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038701.2 NP_001033790.1 8 1109
NM_008071.3 NP_032097.1 8 1161
GenBank mRNA AK039070.1 - 8 1131
AK083051.1 - 8 1173
AK083107.1 - 8 1110
AK140718.1 - 4 704
AK147247.1 - 8 1115
U14420.1 - 8 1142

View Full Product Details