Gene Transcripts

Gene Symbol
Grcc10
Entrez Gene ID
14790
Gene Name
gene rich cluster, C10 gene
Gene Aliases
2310033H05Rik, C10
Chromosome Location
Chr. 6: 124739183 - 124741079 on Build GRCm38
UniGene ID
Mm.439877
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013535.1 NP_038563.1 2 236
GenBank mRNA AK009591.1 - 2 163
BC005683.1 - 2 215 3154555

View Full Product Details