Gene Transcripts

Gene Symbol
Hmx2
Entrez Gene ID
15372
Gene Name
H6 homeobox 2
Gene Aliases
Nkx-5.2, Nkx5-2
Chromosome Location
Chr. 7: 131548773 - 131558867 on Build GRCm38
UniGene ID
Mm.303357
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145998.3 NP_666110.1 2 1001
GenBank mRNA AK053014.1 - 2 771
AK138228.1 - 2 1002
BC023402.1 - 3 871 5043712
S80989.1 - 2 795

View Full Product Details