Gene Transcripts

Gene Symbol
Htr5a
Entrez Gene ID
15563
Gene Name
5-hydroxytryptamine (serotonin) receptor 5A
Gene Aliases
Htr5
Chromosome Location
Chr. 5: 27841702 - 27855086 on Build GRCm38
UniGene ID
Mm.4835
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008314.2 NP_032340.2 1 629
GenBank mRNA AK141636.1 - 1 428
BC115911.1 - 1 321 40104765
BC116654.1 - 1 321 40104769

View Full Product Details