Gene Transcripts

Gene Symbol
Igfals
Entrez Gene ID
16005
Gene Name
insulin-like growth factor binding protein, acid labile subunit
Gene Aliases
ALS, Albs, mKIAA4111
Chromosome Location
Chr. 17: 24877653 - 24882011 on Build GRCm38
UniGene ID
Mm.3135
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008340.3 NP_032366.2 2 1436
GenBank mRNA AK004926.1 - 2 1438
AK220290.1 - 2 1948
BC129875.1 - 2 1424 40110389
BC129876.1 - 2 1424 40110391

View Full Product Details