Gene Transcripts

Gene Symbol
Ipp
Entrez Gene ID
16351
Gene Name
IAP promoted placental gene
Gene Aliases
D4Jhu8, Mipp
Chromosome Location
Chr. 4: 116507511 - 116542561 on Build GRCm38
UniGene ID
Mm.1350
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008389.3 NP_032415.2 7 1386
GenBank mRNA AF285178.1 - 7 1276
AK131718.1 - 7 1387
X58523.1 - 314

View Full Product Details