Gene Transcripts

Gene Symbol
Kcnab2
Entrez Gene ID
16498
Gene Name
potassium voltage-gated channel, shaker-related subfamily, beta member 2
Gene Aliases
F5, I2rf5, Kcnb3, kv-beta-2
Chromosome Location
Chr. 4: 152390740 - 152477549 on Build GRCm38
UniGene ID
Mm.388924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252654.1 NP_001239583.1 5 409
NM_001252655.1 NP_001239584.1 5 438
NM_001252656.1 NP_001239585.1 6 480
NM_010598.3 NP_034728.2 6 451
GenBank mRNA AK143166.1 - 5 386
AK152613.1 - 6 403
AK157053.1 - 5 402
BC039178.1 - 6 381 4503373
L48983.1 - 6 443
U31908.1 - 6 388
U65592.1 - 5 246

View Full Product Details