Gene Transcripts

Gene Symbol
Lhx4
Entrez Gene ID
16872
Gene Name
LIM homeobox protein 4
Gene Aliases
A330062J17Rik, Gsh-4, Gsh4
Chromosome Location
Chr. 1: 155698031 - 155751726 on Build GRCm38
UniGene ID
Mm.103624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010712.2 NP_034842.2 3 565
GenBank mRNA AB221565.1 - 2 346
AF135415.1 - 2 350
BC049834.1 - 3 565 6493203
U89343.1 - 2 328

View Full Product Details