Gene Transcripts

Gene Symbol
Lyz1
Entrez Gene ID
17110
Gene Name
lysozyme 1
Gene Aliases
Lyz, Lzp-s
Chromosome Location
Chr. 10: 117287795 - 117292868 on Build GRCm38
UniGene ID
Mm.177539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013590.4 NP_038618.1 4 1216
GenBank mRNA BC061129.1 - 4 1216

View Full Product Details