Gene Transcripts

Gene Symbol
Mfap2
Entrez Gene ID
17150
Gene Name
microfibrillar-associated protein 2
Gene Aliases
AI893631, Magp, Magp1
Chromosome Location
Chr. 4: 141010424 - 141015984 on Build GRCm38
UniGene ID
Mm.7386
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161799.1 NP_001155271.1 7 423
NM_008546.3 NP_032572.2 7 449
GenBank mRNA AK160213.1 - 7 425
AK164920.1 - 7 406
BC035490.1 - 7 383 5249342
DV654229.1 - 7 423
L23769.1 - 7 405

View Full Product Details